COM448686531 Combi-Blocks, Inc.

Cyclobutanecarboxylic acid, 1-(4-chlorophenyl)-, methyl ester

  • MDL number MFCD25970174
  • Cas Number : 918873-02-6
  • Molecular Weight : 224.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1380
Availability details for
Quantity
  • Availability
  • Add to Cart