COM448686548 Combi-Blocks, Inc.

methyl 2-cyano-1,3-dihydroindene-2-carboxylate

  • MDL number MFCD27578391
  • Cas Number : 260392-88-9
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1412

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1412
Availability details for
Quantity
  • Availability
  • Add to Cart