COM448686550 Combi-Blocks, Inc.

Methyl 3-(2-oxopyrrolidin-1-yl)benzoate

  • MDL number MFCD00593013
  • Cas Number : 329205-78-9
  • Molecular Weight : 219.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1415
Availability details for
Quantity
  • Availability
  • Add to Cart