COM448686552 Combi-Blocks, Inc.

Methyl 1-methyl-5-phenyl-1h-pyrazole-3-carboxylate

  • MDL number MFCD16729663
  • Cas Number : 10250-65-4
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1419

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1419
Availability details for
Quantity
  • Availability
  • Add to Cart