COM448686553 Combi-Blocks, Inc.

methyl 3-(butylsulfamoyl)benzoate

  • MDL number MFCD13262548
  • Cas Number : 1147486-05-2
  • Molecular Weight : 271.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1421
Availability details for
Quantity
  • Availability
  • Add to Cart