COM448686556 Combi-Blocks, Inc.

Methyl 4'-fluoro[1,1'-biphenyl]-3-carboxylate

  • MDL number MFCD05980208
  • Cas Number : 10540-38-2
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1424
Availability details for
Quantity
  • Availability
  • Add to Cart