COM448686557 Combi-Blocks, Inc.

Methyl 2-(4-chlorophenyl)-2-methylpropanoate

  • MDL number MFCD09865030
  • Cas Number : 57225-86-2
  • Molecular Weight : 212.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1427

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1427
Availability details for
Quantity
  • Availability
  • Add to Cart