COM448686560 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-2-cyclopropylacetate

  • MDL number MFCD27578394
  • Cas Number : 1449295-52-6
  • Molecular Weight : 229.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1432

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1432
Availability details for
Quantity
  • Availability
  • Add to Cart