COM448686562 Combi-Blocks, Inc.

Namiki bbv-44227707

  • MDL number MFCD23718302
  • Cas Number : 21024-57-7
  • Molecular Weight : 140.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1437

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1437
Availability details for
Quantity
  • Availability
  • Add to Cart