COM448686563 Combi-Blocks, Inc.

methyl 4-bromo-2-ethoxybenzoate

  • MDL number MFCD27578395
  • Cas Number : 1337607-00-7
  • Molecular Weight : 259.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1438

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1438
Availability details for
Quantity
  • Availability
  • Add to Cart