COM448686564 Combi-Blocks, Inc.

methyl 3-(cyclohexylsulfamoyl)benzoate

  • MDL number MFCD10668696
  • Cas Number : 1088156-10-8
  • Molecular Weight : 297.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1439

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1439
Availability details for
Quantity
  • Availability
  • Add to Cart