COM448686565 Combi-Blocks, Inc.

methyl 5-(2-methylphenyl)-1H-pyrazole-3-carboxylate

  • MDL number MFCD27578396
  • Cas Number : 1239577-96-8
  • Molecular Weight : 216.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1440

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1440
Availability details for
Quantity
  • Availability
  • Add to Cart