COM448686566 Combi-Blocks, Inc.

1-(3,4-Dichloro-phenyl)-cyclopropanecarboxylic acid methyl ester

  • MDL number MFCD18907442
  • Cas Number : 1781330-59-3
  • Molecular Weight : 245.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1442
Availability details for
Quantity
  • Availability
  • Add to Cart