COM448686568 Combi-Blocks, Inc.

Methyl 2,3-difluoro-5-iodobenzoate

  • MDL number MFCD16875976
  • Cas Number : 1163692-49-6
  • Molecular Weight : 298.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1444

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1444
Availability details for
Quantity
  • Availability
  • Add to Cart