COM448686574 Combi-Blocks, Inc.

Methyl 2-(4-(tert-butoxycarbonylamino)phenyl)acetate

  • MDL number MFCD18459353
  • Cas Number : 112918-77-1
  • Molecular Weight : 265.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1456

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1456
Availability details for
Quantity
  • Availability
  • Add to Cart