COM448686576 Combi-Blocks, Inc.

4-Hydroxy-8-bromoquinoline-3-carboxylic acid methyl ester

  • MDL number MFCD22418571
  • Cas Number : 1597073-12-5
  • Molecular Weight : 282.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1463

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1463
Availability details for
Quantity
  • Availability
  • Add to Cart