COM448686577 Combi-Blocks, Inc.

methyl 5-amino-4-chloro-2-fluorobenzoate hydrochloride

  • MDL number MFCD27578399
  • Cas Number : 1820740-71-3
  • Molecular Weight : 240.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1464
Availability details for
Quantity
  • Availability
  • Add to Cart