COM448686578 Combi-Blocks, Inc.

Methyl 3-(pyrrolidine-1-sulfonyl)benzoate

  • MDL number MFCD02949401
  • Cas Number : 494839-04-2
  • Molecular Weight : 269.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1466

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1466
Availability details for
Quantity
  • Availability
  • Add to Cart