COM448686581 Combi-Blocks, Inc.

Boc-D-valine methyl ester

  • MDL number MFCD00672511
  • Cas Number : 106391-85-9
  • Molecular Weight : 231.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. Freezer
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1472
Availability details for
Quantity
  • Availability
  • Add to Cart