COM448686582 Combi-Blocks, Inc.

Methyl cis-(+/-)-3-{[(tert-butoxy)carbonyl]amino}cyclohexane-1-carboxylate

  • MDL number MFCD29035428
  • Cas Number : 930596-35-3
  • Molecular Weight : 257.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1473

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1473
Availability details for
Quantity
  • Availability
  • Add to Cart