COM448686585 Combi-Blocks, Inc.

methyl 5-bromo-2-cyclohexaneamidobenzoate

  • MDL number MFCD04526507
  • Cas Number : 1820683-37-1
  • Molecular Weight : 340.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1478

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1478
Availability details for
Quantity
  • Availability
  • Add to Cart