COM448686588 Combi-Blocks, Inc.

1-(2-Nitro-phenyl)-azetidine-3-carboxylic acid methyl ester

  • MDL number MFCD07787210
  • Cas Number : 887596-00-1
  • Molecular Weight : 236.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1488

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1488
Availability details for
Quantity
  • Availability
  • Add to Cart