COM448686590 Combi-Blocks, Inc.

4-Methyl-benzenepropanoic acid methyl ester

  • MDL number MFCD16663245
  • Cas Number : 56955-36-3
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1493

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1493
Availability details for
Quantity
  • Availability
  • Add to Cart