COM448686593 Combi-Blocks, Inc.

methyl 3-(hexylsulfamoyl)benzoate

  • MDL number MFCD15772994
  • Cas Number : 1820639-13-1
  • Molecular Weight : 299.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1499

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1499
Availability details for
Quantity
  • Availability
  • Add to Cart