COM448686595 Combi-Blocks, Inc.

methyl 7-chloro-4-hydroxy-8-methylquinoline-3-carboxylate

  • MDL number MFCD27578404
  • Cas Number : 1820665-11-9
  • Molecular Weight : 251.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1501

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1501
Availability details for
Quantity
  • Availability
  • Add to Cart