COM448686599 Combi-Blocks, Inc.

methyl 2-methyl-5-oxo-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylate

  • MDL number MFCD27578405
  • Cas Number : 1820740-38-2
  • Molecular Weight : 224.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1513

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1513
Availability details for
Quantity
  • Availability
  • Add to Cart