COM448686600 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(2-methylphenyl)propanoate

  • MDL number MFCD27578406
  • Cas Number : 1201683-61-5
  • Molecular Weight : 293.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1514

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1514
Availability details for
Quantity
  • Availability
  • Add to Cart