COM448686601 Combi-Blocks, Inc.

methyl (2S)-2-{[(tert-butoxy)carbonyl]amino}-5-[3-(2,2,4,6,7-pentamethyl-3H-1-benzofuran-5-sulfonyl)carbamimidamido]pentanoate

  • MDL number MFCD27578407
  • Cas Number : 1820574-57-9
  • Molecular Weight : 540.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Assay 95

Safety Information

msds http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1515
Availability details for
Quantity
  • Availability
  • Add to Cart