COM448686606 Combi-Blocks, Inc.

Methyl 2-[3-methyl-4-(propan-2-yl)phenoxy]acetate

  • MDL number MFCD02283431
  • Cas Number : 649774-36-7
  • Molecular Weight : 222.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1524

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1524
Availability details for
Quantity
  • Availability
  • Add to Cart