COM448686610 Combi-Blocks, Inc.

Methyl 2-(morpholin-4-yl)pyridine-3-carboxylate

  • MDL number MFCD06204161
  • Cas Number : 773134-07-9
  • Molecular Weight : 222.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1529

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1529
Availability details for
Quantity
  • Availability
  • Add to Cart