COM448686613 Combi-Blocks, Inc.

Methyl pyrrolo[1,2-c]pyrimidine-3-carboxylate

  • MDL number MFCD13178547
  • Cas Number : 58298-71-8
  • Molecular Weight : 176.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1533

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1533
Availability details for
Quantity
  • Availability
  • Add to Cart