COM448686617 Combi-Blocks, Inc.

methyl 2-chloro-4-[4-(methoxycarbonyl)phenyl]benzoate

  • MDL number MFCD27578412
  • Cas Number : 161225-09-8
  • Molecular Weight : 304.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1539

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1539
Availability details for
Quantity
  • Availability
  • Add to Cart