COM448686618 Combi-Blocks, Inc.

Methyl 4-(3-pyridinyl)benzoate

  • MDL number MFCD00110082
  • Cas Number : 90395-47-4
  • Molecular Weight : 213.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1540

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1540
Availability details for
Quantity
  • Availability
  • Add to Cart