COM448686620 Combi-Blocks, Inc.

methyl 2-methyl-2-[(2-nitrobenzene)sulfonyl]propanoate

  • MDL number MFCD27578413
  • Cas Number : 1820666-55-4
  • Molecular Weight : 287.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1543

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1543
Availability details for
Quantity
  • Availability
  • Add to Cart