COM448686621 Combi-Blocks, Inc.

methyl 2-methyl-3-[4-(trifluoromethoxy)phenyl]benzoate

  • MDL number MFCD27578414
  • Cas Number : 1630035-51-6
  • Molecular Weight : 310.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1544

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1544
Availability details for
Quantity
  • Availability
  • Add to Cart