COM448686622 Combi-Blocks, Inc.

methyl (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(furan-2-yl)propanoate

  • MDL number MFCD27578415
  • Cas Number : 1820569-92-3
  • Molecular Weight : 391.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1545

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1545
Availability details for
Quantity
  • Availability
  • Add to Cart