COM448686623 Combi-Blocks, Inc.

methyl (2R)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(1,3-thiazol-4-yl)propanoate

  • MDL number MFCD27578416
  • Cas Number : 1820574-14-8
  • Molecular Weight : 408.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1546

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1546
Availability details for
Quantity
  • Availability
  • Add to Cart