COM448686624 Combi-Blocks, Inc.

Methyl 3'-(trifluoromethyl)[1,1'-biphenyl]-4-carboxylate

  • MDL number MFCD05981787
  • Cas Number : 127783-72-6
  • Molecular Weight : 280.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1547

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1547
Availability details for
Quantity
  • Availability
  • Add to Cart