COM448686628 Combi-Blocks, Inc.

methyl (2R)-2-{[(tert-butoxy)carbonyl]amino}-3-(3,4-difluorophenyl)propanoate

  • MDL number MFCD27578418
  • Cas Number : 176795-00-9
  • Molecular Weight : 315.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1552

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1552
Availability details for
Quantity
  • Availability
  • Add to Cart