COM448686634 Combi-Blocks, Inc.

Methyl 4-chlorothieno[3,2-d]pyrimidine-6-carboxylate

  • MDL number MFCD22566148
  • Cas Number : 1363381-65-0
  • Molecular Weight : 228.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1560

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1560
Availability details for
Quantity
  • Availability
  • Add to Cart