COM448686636 Combi-Blocks, Inc.

methyl (2S)-2-acetamido-3-[1-(triphenylmethyl)imidazol-4-yl]propanoate

  • MDL number MFCD27578422
  • Cas Number : 1820580-33-3
  • Molecular Weight : 453.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1563

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1563
Availability details for
Quantity
  • Availability
  • Add to Cart