COM448686641 Combi-Blocks, Inc.

Methyl-3-(4-pyridyl)propanoate

  • MDL number MFCD07367499
  • Cas Number : 90610-07-4
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1568

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1568
Availability details for
Quantity
  • Availability
  • Add to Cart