COM448686642 Combi-Blocks, Inc.

Methyl 3-benzenesulfonamidopropanoate

  • MDL number MFCD14586585
  • Cas Number : 91819-18-0
  • Molecular Weight : 243.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1569

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1569
Availability details for
Quantity
  • Availability
  • Add to Cart