COM448686644 Combi-Blocks, Inc.

1-(9H-fluoren-9-ylmethyl) 2-methyl (2S)-azetidine-1,2-dicarboxylate

  • MDL number MFCD27578426
  • Cas Number : 1820575-03-8
  • Molecular Weight : 337.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1571

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1571
Availability details for
Quantity
  • Availability
  • Add to Cart