COM448686650 Combi-Blocks, Inc.

6-tert-butyl 1-methyl (2S)-2-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}hexanedioate

  • MDL number MFCD27578431
  • Cas Number : 1820574-99-9
  • Molecular Weight : 453.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1578

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1578
Availability details for
Quantity
  • Availability
  • Add to Cart