COM448686651 Combi-Blocks, Inc.

Methyl 6-[3-(trifluoromethyl)phenoxy]-2-pyridinecarboxylate

  • MDL number MFCD00227528
  • Cas Number : 136227-55-9
  • Molecular Weight : 297.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1579

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1579
Availability details for
Quantity
  • Availability
  • Add to Cart