COM448686654 Combi-Blocks, Inc.

methyl (3R)-3-{[(9H-fluoren-9-ylmethoxy)carbonyl]amino}-3-(3-nitrophenyl)propanoate

  • MDL number MFCD27578434
  • Cas Number : 1820574-61-5
  • Molecular Weight : 446.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1582

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1582
Availability details for
Quantity
  • Availability
  • Add to Cart