COM448686655 Combi-Blocks, Inc.

methyl 4-[3,5-bis(trifluoromethyl)phenyl]benzoate

  • MDL number MFCD08701492
  • Cas Number : 198205-83-3
  • Molecular Weight : 348.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1583

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1583
Availability details for
Quantity
  • Availability
  • Add to Cart