COM448686657 Combi-Blocks, Inc.

Methyl benzothiophene-3-carboxylate

  • MDL number MFCD06203855
  • Cas Number : 22913-25-3
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Refrigerated
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1587

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1587
Availability details for
Quantity
  • Availability
  • Add to Cart