COM448686659 Combi-Blocks, Inc.

Methyl 2-amino-3-(3-bromophenyl)propanoate

  • MDL number MFCD19359605
  • Cas Number : 721880-65-5
  • Molecular Weight : 258.11

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1590

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?YC-1590
Availability details for
Quantity
  • Availability
  • Add to Cart